Silicon dioxide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.
Purity: 99.5%
CAS No.: 14808-60-7
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: -325 Mesh Powder
Density: 2.2-2.6
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1710 °C Density: 2.2…2.6 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 238-878-4 Merck Index: 14,8493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...