Zinc sulfide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
88299-09
88299-18
88299-30
CAAA88299-09EA
43.45
CAD
CAAA88299-09
CAAA88299-18
CAAA88299-30
Zinc sulfide ≥99.9% (metals basis)
Zinc sulphide
MDL: MFCD00011301
Purity: 99.9%
CAS No.: 1314-98-3
Molecular Formula: SZn
Formula Weight: 97.43
Physical Form: 3-12mm Pieces
Density: 4.10
Appearance: White
Odor: Odorless
Melting Point: 1700°
Merck Reference: 14,10160
MDL No.: MFCD00011301
Formula:
SZn Melting Pt: 1700 °C Density: 4.10 Storage Temperature: Ambient |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 EINECS: 215-251-3 Merck Index: 14,10160 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...