Lead(II) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
88285-14
88285-22
CAAA88285-14EA
339.7
CAD
CAAA88285-14
CAAA88285-22
Lead(II) telluride ≥99.999% (metals basis)
Lead (II) telluride
MDL: MFCD00016274
Purity: 99.999%
CAS No.: 1314-91-6
Molecular Formula: PbTe
Formula Weight: 334.79
Physical Form: 15mm & Down Pieces
Density: 8.164
Melting Point: 917°
Merck Reference: 14,5422
UN No.: 3284
MDL No.: MFCD00016274
Formula:
PbTe Melting Pt: 917 °C Density: 8.164 Storage Temperature: Ambient |
MDL Number:
MFCD00016274 CAS Number: 1314-91-6 EINECS: 215-247-1 UN: 3077 ADR: 9,III Merck Index: 14,5422 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...