Diindium triselenide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
88280-06
88280-14
CAAA88280-06EA
213.3
CAD
CAAA88280-06
CAAA88280-14
Diindium triselenide ≥99.99% (metals basis)
Diindium triselenide
MDL: MFCD00016147
Purity: 99.99%
CAS No.: 12056-07-4
Molecular Formula: In2Se3
Formula Weight: 466.52
Physical Form: 3-6mm Pieces
Density: 5.67
Melting Point: ca 890°
MDL No.: MFCD00016147
Formula:
In₂Se₃ Melting Pt: ∼ 890 °C Density: 5.67 Storage Temperature: Ambient |
MDL Number:
MFCD00016147 CAS Number: 12056-07-4 EINECS: 235-016-9 UN: 3283 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...