Strontium oxide ≥99.5% (metals basis) SrO ≈97%, powder -100 Mesh

Supplier: Thermo Scientific Chemicals
Danger

88220-06 88220-14 88220-22
CAAA88220-06EA 65.25 CAD
CAAA88220-06 CAAA88220-14 CAAA88220-22
Strontium oxide ≥99.5% (metals basis) SrO ≈97%, powder -100 Mesh
Strontium oxide

Strontium oxide was used to make television picture tube glass. It finds application in glass, optic and ceramic industry. It is an active component of paint dryers. It is also used to prepare pure strontium up on heating with aluminum under vacuum.


Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water.

Formula: SrO
Boiling Pt: ∼3000 °C
Melting Pt: 2430 °C
Density: 4.700
Storage Temperature: Ambient
MDL Number: MFCD00049558
CAS Number: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8,II
Merck Index: 14,8846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR