Magnesium diboride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
88149-06
88149-14
88149-22
CAAA88149-06EA
90.38
CAD
CAAA88149-06
CAAA88149-14
CAAA88149-22
Magnesium diboride ≥99% (metals basis)
Magnesium diboride
MDL: MFCD00049752
Purity: 99%
CAS No.: 12007-25-9
Molecular Formula: B2Mg
Formula Weight: 45.93
Physical Form: -100 Mesh Powder
Appearance: Dark grey
Odor: Odorless
Melting Point: 800° dec.
MDL No.: MFCD00049752
Formula:
B₂Mg Melting Pt: 800 °C (dec.) Density: 2,3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049752 CAS Number: 12007-25-9 EINECS: 234-501-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...