Terbium, AAS Standard Solution, Specpure®, Tb 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
88110-AP 88110-AE 88110-AP 88110-AE
CAAA88110-APEA 162.74 CAD
CAAA88110-AP CAAA88110-AE AA88110-AP AA88110-AE
Terbium, AAS Standard Solution, Specpure®, Tb 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Terbium Standards
MDL: MFCD00011256

Molecular Formula: Tb4O7 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011256
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR