Chlorosulfuric acid ∼99%

Supplier: Thermo Scientific Chemicals
Danger

87977-30 87977-A1 87977-D5
CAAA87977-30EA 138.72 CAD
CAAA87977-30 CAAA87977-A1 CAAA87977-D5
Chlorosulfuric acid ∼99%
Chlorosulphuric acid

MDL: MFCD00011523


Purity: 99% CAS No.: 7790-94-5 Molecular Formula: HSO3Cl Formula Weight: 116.52 Physical Form: Liquid Density: 1.753 Refractive Index: 1.433 Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Odor: Pungent Boiling Point: 158° Merck Reference: 14,2165 UN No.: 1754 MDL No.: MFCD00011523

Formula: HClO₃S
Boiling Pt: 158 °C
Melting Pt: –80 °C
Density: 1.753
Storage Temperature: Ambient
MDL Number: MFCD00011523
CAS Number: 7790-94-5
EINECS: 232-234-6
UN: 1754
ADR: 8,I
Merck Index: 14,2165

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR