Perchloric acid 69,0 - 72,0% ACS, redistilled
Supplier: Thermo Scientific Chemicals
|
Danger
|
87963-AP
87963-K4
CAAA87963-APEA
243.32
CAD
CAAA87963-AP
CAAA87963-K4
Perchloric acid 69,0 - 72,0% ACS, redistilled
Perchloric acid
MDL: MFCD00011325
Fieser: 1,796 2,309 3,220 5,506 6,453 7,279 8,386 11,402 12,402
Purity: 70%
CAS No.: 7601-90-3
Molecular Formula: HClO4
Formula Weight: 100.46
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.67
Appearance: Colorless
Odor: Odorless
Melting Point: -112°
Boiling Point: 203°
Merck Reference: 14,7153
UN No.: 1873
MDL No.: MFCD00011325
Formula:
HClO₄ Boiling Pt: 203 °C Melting Pt: –112 °C Density: 1.67 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 1873 ADR: 5.1,I Merck Index: 14,7153 |
|
Specification Test Results
Color (APHA) | 10 |
Residue after ignition | 0.003 |
Silicate and phosphate (as SiO2 | 5 ppm |
Cl | 0.001% |
Nitrogen compounds (as N) | 0.001% |
SO3 | 0.001% |
Heavy Metals (as Pb) | 1 ppm |
Fe | 1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...