Nitric acid 65 - 70% ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

87920-AE 87920-AP
CAAA87920-AEEA 94.8 CAD
CAAA87920-AE CAAA87920-AP
Nitric acid 65 - 70% ≥99.999% (metals basis)
Nitric acid

In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuels


Purity: 99.999% CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Density: 1.41 Appearance: Colorless to pale yellow Odor: Pungent Boiling Point: 121° Solubility: Very soluble in water and ether. Decomposes in alcohol Merck Reference: 14,6577 UN No.: 2031 MDL No.: MFCD00011349


In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuels

Formula: HNO₃
Boiling Pt: 121 °C
Melting Pt: –42 °C
Density: 1.41
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 14,6577

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR