Thallium(III) trifluoroacetate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thallic trifluoroacetate
87913-04
87913-09
87913-18
CAAA87913-04EA
75.68
CAD
CAAA87913-04
CAAA87913-09
CAAA87913-18
Thallium(III) trifluoroacetate ≥95%
Thallium (III) trifluoroacetate
MDL: MFCD00000414
Purity: 95%
CAS No.: 23586-53-0
Molecular Formula: Tl(OOCCF3)3
Formula Weight: 543.42
Physical Form: Solid
Sensitivity: Hygroscopic
UN No.: 1707
MDL No.: MFCD00000414
Formula:
C₆F₉O₆Tl Boiling Pt: 160 °C (1.5 mmHg) Melting Pt: 213 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00000414 CAS Number: 23586-53-0 EINECS: 245-761-1 UN: 1707 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...