Ammonia solution 25% ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
87903-AK
87903-AU
CAAA87903-AKEA
230.68
CAD
CAAA87903-AK
CAAA87903-AU
Ammonia solution 25% ≥99.99% (metals basis)
Ammonia solution
MDL: MFCD00066650
Purity: 99.99%
CAS No.: 1336-21-6
Molecular Formula: H5NO
Formula Weight: 35.05
Physical Form: Liquid
Density: 0.91
Appearance: Colorless
Odor: Ammonia-like
pH: 12
Merck Reference: 14,494
UN No.: 2672
MDL No.: MFCD00066650
Formula:
NH₃ Boiling Pt: 32…37.7 °C (1013 hPa) Melting Pt: –60…–57.5 °C Density: 0.91 Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III Merck Index: 14,494 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...