Calcium bromide, anhydrous ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Photography, dehydrating agent, manufacturing of mineral water, fire retardant, wood preservative, freezing mixture, road treatment
Purity: 99.5%
CAS No.: 7789-41-5
Molecular Formula: CaBr2
Formula Weight: 199.90
Physical Form: -50 Mesh Powder
Density: 3.353
Appearance: White to off-white
Odor: Odorless
Melting Point: 730°
Boiling Point: 806-812°
Solubility: Freely soluble in water, methanol, ethanol. Soluble in acetone. Insoluble in dioxane, chloroform, ether
Merck Reference: 14,1654
MDL No.: MFCD00010902
Photography, dehydrating agent, manufacturing of mineral water, fire retardant, wood preservative, freezing mixture, road treatment
Formula:
Br₂Ca MW: 199.89 g/mol Boiling Pt: 810 Melting Pt: 730 Density: 3.353 Storage Temperature: Ambient |
MDL Number:
MFCD00010902 CAS Number: 7789-41-5 EINECS: 232-164-6 Merck Index: 14,1654 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...