Sodium cyanotrihydroborate ≥95%

Supplier: Thermo Scientific Chemicals
Danger

87839-06 87839-14 87839-22
CAAA87839-06EA 57.35 CAD
CAAA87839-06 CAAA87839-14 CAAA87839-22
Sodium cyanotrihydroborate ≥95%
Sodium cyanotrihydroborate

Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines


Purity: 95% CAS No.: 25895-60-7 Molecular Formula: NaBH3CN Formula Weight: 62.84 Physical Form: Powder Density: 1.20 Melting Point: 300° dec. Solubility: Soluble in water, THF, methanol Merck Reference: 14,8606 UN No.: 3134 MDL No.: MFCD00003516


Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines

Formula: CH₃BNNa
Melting Pt: >300 °C (dec.)
Density: 1.20
Storage Temperature: Ambient
MDL Number: MFCD00003516
CAS Number: 25895-60-7
EINECS: 247-317-2
UN: 3134
ADR: 4.3,II
Merck Index: 14,8606

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR