Tellurium dioxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
87813-14
87813-22
CAAA87813-14EA
186.44
CAD
CAAA87813-14
CAAA87813-22
Tellurium dioxide ≥99.99% (metals basis)
Tellurium dioxide
MDL: MFCD00011263
Purity: 99.99%
CAS No.: 7446-07-3
Molecular Formula: TeO2
Formula Weight: 159.60
Physical Form: Powder
Density: 5.8
Melting Point: 733°
Merck Reference: 14,9123
MDL No.: MFCD00011263
Formula:
O₂Te MW: 159.6 g/mol Boiling Pt: 1245 °C (1013 hPa) Melting Pt: 733 °C Density: 5.8 Storage Temperature: Ambient |
MDL Number:
MFCD00011263 CAS Number: 7446-07-3 EINECS: 231-193-1 Merck Index: 14,9123 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...