Tellurium tetraiodide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
87795-06
87795-14
CAAA87795-06EA
115.02
CAD
CAAA87795-06
CAAA87795-14
Tellurium tetraiodide ≥99% (metals basis)
Tellurium tetraiodide
MDL: MFCD00049578
Decomposes slowly in cold water to TeO2 and HI
Purity: 99%
CAS No.: 7790-48-9
Molecular Formula: TeI4
Formula Weight: 635.22
Physical Form: Crystalline
Density: 5.403
Melting Point: 280°
Boiling Point: dec.
Solubility: Decomposes slowly in cold water to TeO2 and HI
Merck Reference: 14,9127
UN No.: 2923
MDL No.: MFCD00049578
Formula:
I₄Te Boiling Pt: Dec. Melting Pt: 280 °C Density: 5.403 Storage Temperature: Ambient |
MDL Number:
MFCD00049578 CAS Number: 7790-48-9 EINECS: 232-210-5 UN: 3260 ADR: 8,II Merck Index: 14,9127 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...