Manganese(III) oxide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
87791-36
87791-A3
CAAA87791-36EA
281.24
CAD
CAAA87791-36
CAAA87791-A3
Manganese(III) oxide 98%
Manganese (III) oxide
MDL: MFCD00016217
Purity: 98%
CAS No.: 1317-34-6
Molecular Formula: Mn2O3
Formula Weight: 157.86
Physical Form: -325 Mesh Powder
Density: 4.50
Appearance: Black
Odor: Odorless
Melting Point: 1080° dec.
Merck Reference: 14,5737
MDL No.: MFCD00016217
Formula:
Mn₂O₃ Melting Pt: 1080 °C (dec.) Density: 4.50 Storage Temperature: Ambient |
MDL Number:
MFCD00016217 CAS Number: 1317-34-6 EINECS: 215-264-4 Merck Index: 14,5737 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...