Iodine(V) oxide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
di-Iodine pentaoxide, Iodopentoxide, di-Iodine pentoxide
An oxidizing agent to remove CO from gas
Purity: 98%
CAS No.: 12029-98-0
Molecular Formula: I2O5
Formula Weight: 333.81
Physical Form: Crystalline
Density: 5.08
Appearance: White to cream
pH: 1.1
Melting Point: 275-350° dec.
Solubility: Soluble in water and HNO3. Insoluble in absolute alcohol, chloroform, ether, and CS2
Merck Reference: 14,5019
UN No.: 3085
MDL No.: MFCD00011356
An oxidizing agent to remove CO from gas
Formula:
I₂O₅ MW: 333.81 g/mol Melting Pt: 275…350 °C (dec.) Density: 5.08 Storage Temperature: Ambient |
MDL Number:
MFCD00011356 CAS Number: 12029-98-0 EINECS: 234-740-2 UN: 3085 ADR: 5.1 II Merck Index: 14,5019 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...