Antimony triacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
87657-09
87657-22
87657-36
CAAA87657-09EA
41.55
CAD
CAAA87657-09
CAAA87657-22
CAAA87657-36
Antimony triacetate 97%
Antimony triacetate
MDL: MFCD00014974
Decomposes in water
Purity: 97%
CAS No.: 6923-52-0
Molecular Formula: Sb(OOCCH3)3
Formula Weight: 298.88
Physical Form: Powder
Sensitivity: Moisture Sensitive
Solubility: Decomposes in water
UN No.: 1549
MDL No.: MFCD00014974
Formula:
(H₃CCOO)₃Sb Storage Temperature: Ambient |
MDL Number:
MFCD00014974 CAS Number: 6923-52-0 EINECS: 230-043-2 UN: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...