Cesium bromide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cesiumbromide

87639-09 87639-18
CAAA87639-09EA 195.92 CAD
CAAA87639-09 CAAA87639-18
Cesium bromide ≥99.999% (metals basis)
Cesium bromide

MDL: MFCD00010954


Purity: 99.999% CAS No.: 7787-69-1 Molecular Formula: BrCs Formula Weight: 212.81 Physical Form: -4 Mesh Density: 4.44 Refractive Index: 1.6984 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 636° Boiling Point: 1300° Merck Reference: 14,2009 MDL No.: MFCD00010954

Formula: BrCs
MW: 212.81 g/mol
Boiling Pt: 1300 °C
Melting Pt: 636 °C
Density: 4.44
Storage Temperature: Ambient
MDL Number: MFCD00010954
CAS Number: 7787-69-1
EINECS: 232-130-0
Merck Index: 14,2009

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR