Potassium chloride ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
87626-18
87626-30
CAAA87626-18EA
210.14
CAD
CAAA87626-18
CAAA87626-30
Potassium chloride ≥99.995% (metals basis)
Potassium chloride
MDL: MFCD00011360
Fieser: 6,480
Purity: 99.995%
CAS No.: 7447-40-7
Molecular Formula: KCl
Formula Weight: 74.55
Physical Form: Powder
Density: 1.984
Refractive Index: 1.490
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 773°
Boiling Point: 1500° subl.
Merck Reference: 14,7621
MDL No.: MFCD00011360
Formula:
KCl Boiling Pt: 1500 °C (subl.) Melting Pt: 773 °C Density: 1.984 Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,7621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...