Sodium sulfate, anhydrous ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
87611-06
87611-14
87611-22
CAAA87611-06EA
88.16
CAD
CAAA87611-06
CAAA87611-14
CAAA87611-22
Sodium sulfate, anhydrous ≥99.99% (metals basis)
Sodium sulphate
MDL: MFCD00003504
Fieser: 1,1103
Purity: 99.99%
CAS No.: 7757-82-6
Molecular Formula: Na2O4S
Formula Weight: 142.04
Physical Form: Powder/chunks
Density: 2.680
Refractive Index: 1.4840
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 884°
Merck Reference: 14,8680
MDL No.: MFCD00003504
Formula:
Na₂SO₄ Boiling Pt: 1429 °C (1013 hPa) Melting Pt: 884 °C Density: 2.680 Storage Temperature: Ambient |
MDL Number:
MFCD00003504 CAS Number: 7757-82-6 EINECS: 231-820-9 Merck Index: 14,8680 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...