Sodium bromide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
87604-06
87604-14
CAAA87604-06EA
99.54
CAD
CAAA87604-06
CAAA87604-14
Sodium bromide ≥99.99% (metals basis)
Sodium bromide
MDL: MFCD00003475
Fieser: 12,445
Purity: 99.99%
CAS No.: 7647-15-6
Molecular Formula: NaBr
Formula Weight: 102.89
Physical Form: Crystalline
Density: 3.203
Refractive Index: 1.6412
Sensitivity: Hygroscopic
Melting Point: 755°
Boiling Point: 1390°
Merck Reference: 14,8594
MDL No.: MFCD00003475
Formula:
NaBr Boiling Pt: 1390 °C Melting Pt: 755 °C Density: 3.203 Storage Temperature: Ambient |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 Merck Index: 14,8594 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...