Sodium hydrogensulfide, anhydrous

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium hydrogen sulfide, Sodium hydrogen sulphide

87389-04 87389-09
CAAA87389-04EA 311.26 CAD
CAAA87389-04 CAAA87389-09
Sodium hydrogensulfide, anhydrous
Sodium hydrogensulphide

In paper pulping, in dyestuffs processing, in rayon and cellophane desulfurizing, in unhairing hides and as a bleaching reagent


CAS No.: 16721-80-5 Molecular Formula: NaHS, may contain other sulfide phases Formula Weight: 56.06 Physical Form: Crystalline Density: 1.79 Sensitivity: Moisture Sensitive and Hygroscopic Appearance: Off-white Odor: Like rotten eggs Melting Point: 350° UN No.: 2318 MDL No.: MFCD00011124


In paper pulping, in dyestuffs processing, in rayon and cellophane desulfurizing, in unhairing hides and as a bleaching reagent

Formula: HNaS
MW: 56.06 g/mol
Melting Pt: 350 °C
Density: 1.79
Storage Temperature: Ambient
MDL Number: MFCD00011124
CAS Number: 16721-80-5
EINECS: 240-778-0
UN: 2318
ADR: 4.2,II
Merck Index: 12,08730

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR