Cobalt(II) benzoate
Supplier: Thermo Scientific Chemicals
|
Warning
|
87380-09
87380-18
CAAA87380-09EA
63.67
CAD
CAAA87380-09
CAAA87380-18
CAAA87380-30
Cobalt(II) benzoate
Cobalt(II) benzoate
MDL: MFCD00050769
CAS No.: 932-69-4
Molecular Formula: C14H10CoO4
Formula Weight: 301.16
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Grey
Odor: Odorless
Melting Point: 114-116°
MDL No.: MFCD00050769
Formula:
C₁₄H₁₀CoO₄ Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050769 CAS Number: 932-69-4 EINECS: 213-257-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...