Sodium tungstate dihydrate ≥95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate

87371-30 87371-A1
CAAA87371-30EA 152.94 CAD
CAAA87371-30 CAAA87371-A1
Sodium tungstate dihydrate ≥95%
Sodium wolframate dihydrate

MDL: MFCD00149190 Soluble in water. Insoluble in alcohol Fieser: 13,145 15,295


Purity: 95% CAS No.: 10213-10-2 Molecular Formula: Na2WO4.2H2O Formula Weight: 329.86 (293.83anhy) Physical Form: Crystalline Density: 3.250 Appearance: White Odor: Odorless Melting Point: 100° -2H2O Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,8698 MDL No.: MFCD00149190

Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Melting Pt: 100
Density: 3.250
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
EINECS: 236-743-4
Merck Index: 14,8698

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR