Silver chloride ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Silver plating
Purity: 95%
CAS No.: 7783-90-6
Molecular Formula: AgCl
Formula Weight: 143.32
Physical Form: Coarse Powder
Density: 5.56
Refractive Index: 2.071
Sensitivity: Light Sensitive
Appearance: White
Odor: Odorless
Melting Point: 455°
Boiling Point: 1550°
Solubility: Practically insoluble in water. Soluble in ammonia, solutions of alkali cyanides, thiosulfates, ammonium carbonates. Insoluble in alcohol, dilute acids
Merck Reference: 14,8509
UN No.: 3077
MDL No.: MFCD00003399
Silver plating
Formula:
AgCl Boiling Pt: 1550 °C Melting Pt: 455 °C Density: 5.56 Storage Temperature: Ambient |
MDL Number:
MFCD00003399 CAS Number: 7783-90-6 EINECS: 232-033-3 UN: 3077 ADR: 9,III Merck Index: 14,8509 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...