Nickel (II) oxide, black Ni_76 76% Ni (contains Ni2O3)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel monoxide, Nickel oxide, Bunsenite

87302-30 87302-A1
CAAA87302-30EA 136.67 CAD
CAAA87302-30 CAAA87302-A1
Nickel (II) oxide, black Ni_76 76% Ni (contains Ni2O3)
Nickel (II) oxide
Formula: NiO
MW: 74.69 g/mol
Melting Pt: 1990
Density: 6.67
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7
Merck Index: 14,6512

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR