Mercury(I) chloride ≥99.5%

Supplier: Thermo Scientific Chemicals
Warning

87240-22 87240-36
CAAA87240-22EA 120.55 CAD
CAAA87240-22 CAAA87240-36
Mercury(I) chloride ≥99.5%
Mercury (I) chloride

For calomel electrodes, in ceramic painting, porcelain painting with gold

Formula: Cl₂Hg₂
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 400 °C (subl.)
Density: 7.15
Storage Temperature: Ambient
MDL Number: MFCD00011043
CAS Number: 10112-91-1
EINECS: 233-307-5
UN: 2025
ADR: 6.1,II
Merck Index: 14,5894

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR