Boron triiodide ≥95%

Supplier: Thermo Scientific Chemicals
Danger

87220-03 87220-06
CAAA87220-03EA 186.44 CAD
CAAA87220-03 CAAA87220-06
Boron triiodide ≥95%
Boron triiodide

MDL: MFCD00036286


Purity: 95% CAS No.: 13517-10-7 Molecular Formula: BI3 Formula Weight: 391.52 Physical Form: Crystalline Density: 3.35 Appearance: White to pink Odor: Pungent Melting Point: 48-51° Boiling Point: 210° UN No.: 3260 MDL No.: MFCD00036286

Formula: BI₃
Boiling Pt: 210 °C
Melting Pt: 48…51 °C
Density: 3.35
Storage Temperature: Ambient
MDL Number: MFCD00036286
CAS Number: 13517-10-7
EINECS: 236-857-4
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR