Ferrocene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
87202-18
87202-30
CAAA87202-18EA
60.2
CAD
CAAA87202-18
CAAA87202-30
Ferrocene 99%
Ferrocene
MDL: MFCD00001427
Purity: 99%
CAS No.: 102-54-5
Molecular Formula: C10H10Fe
Formula Weight: 186.04
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Yellow to orange
Odor: Characteristic
Melting Point: 173-174°
Boiling Point: 249°
Merck Reference: 14,4037
UN No.: 1325
MDL No.: MFCD00001427
Formula:
C₁₀H₁₀Fe Boiling Pt: 249 °C Melting Pt: 173…174 °C Density: 1.49 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001427 CAS Number: 102-54-5 EINECS: 203-039-3 UN: 1325 ADR: 4.1,II Merck Index: 14,4037 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...