Zirconium tetrabromide 99%

Supplier: Thermo Scientific Chemicals
Danger

86139-09 86139-18
CAAA86139-09EA 186.44 CAD
CAAA86139-09 CAAA86139-18
Zirconium tetrabromide 99%
Zirconium tetrabromide

MDL: MFCD00049637


Purity: 99% CAS No.: 13777-25-8 Molecular Formula: ZrBr4 Formula Weight: 410.86 Physical Form: -40 Mesh Powder Density: 4.201 Melting Point: 450°/15 atm UN No.: 3260 MDL No.: MFCD00049637

Formula: Br₄Zr
MW: 410.84 g/mol
Melting Pt: 450
Density: 4.201
Storage Temperature: Ambient
MDL Number: MFCD00049637
CAS Number: 13777-25-8
EINECS: 237-417-4
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR