Zirconyl chloride hydrate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zirconium(IV) dichloride oxide hydrate, Zirconium dichloride oxide hydrate
86108-18
86108-30
86108-A1
CAAA86108-18EA
95.75
CAD
CAAA86108-18
CAAA86108-30
CAAA86108-A1
Zirconyl chloride hydrate ≥99.9% (metals basis)
Zirconyl chloride hydrate
MDL: MFCD00149898
Soluble in water, alcohol
Purity: 99.9%
CAS No.: 13520-92-8
Molecular Formula: ZrOCl2.8H2O
Formula Weight: 322.25 (178.13anhy)
Physical Form: Lump
Density: 1.91
Appearance: White
Odor: Acidic
Melting Point: 400° dec.
Solubility: Soluble in water, alcohol
Merck Reference: 14,10184
UN No.: 3260
MDL No.: MFCD00149898
Formula:
Cl₂OZr·H₂O MW: 196.14 g/mol Melting Pt: 400 °C (dec.) Density: 1.91 Storage Temperature: Ambient |
MDL Number:
MFCD00149897 CAS Number: 15461-27-5 EINECS: 231-717-9 UN: 3260 ADR: 8,II Merck Index: 14,10184 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...