Tungstic(VI) acid

Supplier: Thermo Scientific Chemicals
Warning

82118-14 82118-22 82118-36
CAAA82118-14EA 51.51 CAD
CAAA82118-14 CAAA82118-22 CAAA82118-36
Tungstic(VI) acid
Tungstic (VI) acid

MDL: MFCD00011336


CAS No.: 7783-03-1 Molecular Formula: H2WO4 Formula Weight: 249.86 Physical Form: Powder Density: 5.5 Refractive Index: 2.24 Melting Point: 100° dec. Boiling Point: 1473° Merck Reference: 14,9818 MDL No.: MFCD00011336

Formula: H₂O₄W
Boiling Pt: 1473 °C
Melting Pt: 100 °C (dec.)
Density: 5.5
Storage Temperature: Ambient
MDL Number: MFCD00011336
CAS Number: 7783-03-1
EINECS: 231-975-2
Merck Index: 14,9818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR