Vanadium(III) bromide ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
81135-04
81135-09
81135-18
CAAA81135-04EA
134.77
CAD
CAAA81135-04
CAAA81135-09
CAAA81135-18
Vanadium(III) bromide ≥99.5% (metals basis)
Vanadium (III) bromide
MDL: MFCD00049695
Purity: 99.5%
CAS No.: 13470-26-3
Molecular Formula: VBr3
Formula Weight: 290.67
Physical Form: Crystalline
Density: 4.00
Melting Point: dec.
UN No.: 2923
MDL No.: MFCD00049695
Formula:
Br₃V Melting Pt: Dec. Density: 4.00 Storage Temperature: Ambient |
MDL Number:
MFCD00049695 CAS Number: 13470-26-3 EINECS: 236-736-6 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...