Vanadium oxytrichloride V⁺⁵ ≥ 28,5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Vanadium oxychloride, Vanadium (V) oxide chloride, Vanadium (V) trichloride oxide, Vanadyl trichloride, Trichlorooxovanadium, Vanadium (V) chloride, Vanadium trichloride oxide
MDL: MFCD00011459
CAS No.: 7727-18-6
Molecular Formula: VOCl3
Formula Weight: 173.30
Physical Form: Liquid
Density: 1.829
Sensitivity: Moisture Sensitive
Appearance: Yellow
Odor: Chlorine-like
Melting Point: -77°
Boiling Point: 126-127°
Merck Reference: 14,9927
UN No.: 2443
MDL No.: MFCD00011459
Formula:
Cl₃OV MW: 173.3 Boiling Pt: 127 Melting Pt: -77.2 Density: 1.829 Storage Temperature: Ambient |
MDL Number:
MFCD00011459 CAS Number: 7727-18-6 EINECS: 231-780-2 UN: 2443 ADR: 8 II Merck Index: 14,9927 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...