Vanadium(III) fluoride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
81115-04
81115-09
81115-18
CAAA81115-04EA
237
CAD
CAAA81115-04
CAAA81115-09
CAAA81115-18
Vanadium(III) fluoride 98%
Vanadium (III) fluoride
MDL: MFCD00042553
Practically insoluble in water, acetone, and glacial acetic acid
Purity: 98%
CAS No.: 10049-12-4
Molecular Formula: VF3
Formula Weight: 107.94
Physical Form: Powder
Density: 3.363
Melting Point: ≈1406°
Solubility: Practically insoluble in water, acetone, and glacial acetic acid
Merck Reference: 14,9922
UN No.: 2923
MDL No.: MFCD00042553
Formula:
F₃V Melting Pt: ∼1406 °C Density: 3.363 Storage Temperature: Ambient |
MDL Number:
MFCD00042553 CAS Number: 10049-12-4 EINECS: 233-169-6 UN: 2923 ADR: 8,II Merck Index: 14,9922 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...