Sodium orthovanadate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
81104-14
81104-22
81104-36
CAAA81104-14EA
143.62
CAD
CAAA81104-14
CAAA81104-22
CAAA81104-36
Sodium orthovanadate ≥99.9% (metals basis)
Sodium orthovanadate
Inhibitor of protein tyrosine phosphatase and alkaline phosphate
Purity: 99.9%
CAS No.: 13721-39-6
Molecular Formula: Na3O4V
Formula Weight: 183.94
Physical Form: Granular
Appearance: White
Melting Point: 850-866°
MDL No.: MFCD00003511
Inhibitor of protein tyrosine phosphatase and alkaline phosphate
Formula:
Na₃O₄V MW: 183.91 g/mol Melting Pt: 850…866 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003511 CAS Number: 13721-39-6 EINECS: 237-287-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...