Thallium(I) fluoride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
78111-14
78111-22
CAAA78111-14EA
322.32
CAD
CAAA78111-14
CAAA78111-22
Thallium(I) fluoride 97%
Thallium (I) fluoride
Purity: 97%
CAS No.: 7789-27-7
Molecular Formula: TlF
Formula Weight: 223.37
Physical Form: Crystalline
Density: 8.36
Boiling Point: 655°
Solubility: Very soluble in water
Merck Reference: 14,9262
UN No.: 1707
MDL No.: MFCD00049607
Formula:
FTl MW: 223.38 Boiling Pt: 645 Melting Pt: 315 Density: 8.36 Storage Temperature: Ambient |
MDL Number:
MFCD00049607 CAS Number: 7789-27-7 EINECS: 232-154-1 UN: 1707 ADR: 6.1 II Merck Index: 14,9262 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...