Titanium(IV) ethanolate TiO₂ ≥ 33%

Supplier: Thermo Scientific Chemicals
Warning

77142-06 77142-18 77142-30
CAAA77142-06EA 58.46 CAD
CAAA77142-06 CAAA77142-18 CAAA77142-30
Titanium(IV) ethanolate TiO₂ ≥ 33%
Titanium(IV) ethanolate

CAS No.: 3087-36-3 Molecular Formula: C8H20O4Ti Formula Weight: 228.15 Flash Point: 28°(84°F) Physical Form: Liquid Density: 1.088 Refractive Index: 1.5043 Sensitivity: Moisture Sensitive Appearance: Pale yellow Odor: Alcohol-like Melting Point: -40° Boiling Point: 150-152°/10mm Solubility: Soluble in organic solvents. Decomposes in water UN No.: 2924 MDL No.: MFCD00009071

Formula: C₈H₂₀O₄Ti
Boiling Pt: 150…152 °C (10 mmHg)
Melting Pt: –40 °C
Density: 1.088
Flash Pt: 28 °C (84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009071
CAS Number: 3087-36-3
EINECS: 221-410-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR