Titanium(IV) methoxide ≥95%

Supplier: Thermo Scientific Chemicals
Warning

77137-14 77137-22 77137-36
CAAA77137-14EA 142.99 CAD
CAAA77137-14 CAAA77137-22 CAAA77137-36
Titanium(IV) methoxide ≥95%
Titanium (IV) methoxide

Purity: 95% CAS No.: 992-92-7 Molecular Formula: Ti(OCH3)4.(CH3OH)x, x1 Flash Point: 48°(118°F) Physical Form: Granular/ lump Sensitivity: Moisture Sensitive Boiling Point: 243°/52mm UN No.: 3181 MDL No.: MFCD00014883

Formula: C₄H₁₂O₄Ti
Boiling Pt: 243 °C (52 mmHg)
Melting Pt: 200…210 °C
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014883
CAS Number: 992-92-7
EINECS: 213-596-4
UN: 3181
ADR: 4.1,III

Order Now

Specification Test Results

Form Granular/ lump

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR