Titanium(II) oxide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Titanium oxide, Titanium monoxide

77126-06 77126-14 77126-22 77126-36
CAAA77126-06EA 52.61 CAD
CAAA77126-06 CAAA77126-14 CAAA77126-22 CAAA77126-36
Titanium(II) oxide ≥99.5% (metals basis)
Titanium (II) oxide

Purity: 99.5% CAS No.: 12137-20-1 Molecular Formula: TiO Formula Weight: 63.90 Physical Form: -325 Mesh Powder Density: 4.95 Melting Point: 1700° Boiling Point: 3000° MDL No.: MFCD00036279

Formula: OTi
MW: 63.88
Boiling Pt: 3660
Melting Pt: 1750
Density: 4.95
Storage Temperature: Ambient
MDL Number: MFCD00036279
CAS Number: 12137-20-1
EINECS: 235-236-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR