Titanium(IV) isopropoxide 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids
Purity: 97+%
CAS No.: 546-68-9
Molecular Formula: C12H28O4Ti
Formula Weight: 284.23
Flash Point: 46°(115°F)
Physical Form: Liquid
Density: 0.955
Refractive Index: 1.4640
Sensitivity: Moisture Sensitive
Appearance: Colorless to pale yellow
Melting Point: 16-20°
Boiling Point: 232°
Solubility: Soluble in anhydrous ethanol, ether, benzene and chloroform. Decomposes in water
Beilstein Reference: 3679474
Merck Reference: 14,9480
UN No.: 2413
MDL No.: MFCD00008871
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids
Formula:
C₁₂H₂₈O₄Ti Boiling Pt: 232 °C Melting Pt: 16…20 °C Density: 0.955 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008871 CAS Number: 546-68-9 EINECS: 208-909-6 UN: 2413 ADR: 3,III Merck Index: 14,9480 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...