Sodium tellurate(VI) hydrate Te ≥ 45% ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
75106-09
75106-18
CAAA75106-09EA
117.24
CAD
CAAA75106-09
CAAA75106-18
Sodium tellurate(VI) hydrate Te ≥ 45% ≥95%
di-Sodium tellurium tetraoxide hydrate
Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.
CAS No.: 10101-83-4
Molecular Formula: Na2TeO4.xH2O
Formula Weight: 237.58(anhy)
Physical Form: Crystalline
Melting Point: dec.
Merck Reference: 14,8686
MDL No.: MFCD00014242
Formula:
Na₂O₄Te·H₂O Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00014242 CAS Number: 10101-83-4 EINECS: 233-259-5 UN: 3284 ADR: 6.1,II Merck Index: 14,8686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...