Tellurium tetrabromide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tellurium(IV) bromide
75101-09
75101-18
CAAA75101-09EA
191.18
CAD
CAAA75101-09
CAAA75101-18
Tellurium tetrabromide ≥99.9% (metals basis)
Tellurium tetrabromide
MDL: MFCD00049575
Decomposes in water forming TeO2 and HBr
Purity: 99.9%
CAS No.: 10031-27-3
Molecular Formula: TeBr4
Formula Weight: 447.27
Physical Form: -4 Mesh Crystalline
Density: 4.31
Melting Point: 380°
Boiling Point: 420° dec.
Solubility: Decomposes in water forming TeO2 and HBr
Merck Reference: 14,9125
UN No.: 2923
MDL No.: MFCD00049575
Formula:
Br₄Te Boiling Pt: 420 °C (dec.) Melting Pt: 380 °C Density: 4.31 Storage Temperature: Ambient |
MDL Number:
MFCD00049575 CAS Number: 10031-27-3 EINECS: 233-090-7 UN: 2923 ADR: 8,II Merck Index: 14,9125 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...