Terbium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

74103-06 74103-14
CAAA74103-06EA 118.5 CAD
CAAA74103-06 CAAA74103-14
Terbium(III) nitrate hydrate ≥99.9% (REO, rare earth oxide basis)
Terbium(III) nitrate hydrate

MDL: MFCD00011257


Purity: 99.9% CAS No.: 94219-56-4 Molecular Formula: Tb(NO3)3.xH2O Formula Weight: 344.94(anhy) Physical Form: Wet Crystalline Aggregates, Packaged under argon Appearance: White Merck Reference: 14,9157 UN No.: 1477 MDL No.: MFCD00011257

Formula: Tb(NO₃)₃·xH₂O
Melting Pt: 89.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00011257
CAS Number: 94219-56-4
EINECS: 233-138-7
UN: 1477
ADR: 5.1,III
Merck Index: 14,9157

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR