Tantalum(V) oxide ≥99.5% (metals basis), Spectrographic Grade

Supplier: Thermo Scientific Chemicals

Synonyms: Ditantalum pentaoxide

73118-14 73118-22
CAAA73118-14EA 192.76 CAD
CAAA73118-14 CAAA73118-22
Tantalum(V) oxide ≥99.5% (metals basis), Spectrographic Grade
Tantalum (V) oxide

MDL: MFCD00011254


Purity: 99.5% CAS No.: 1314-61-0 Molecular Formula: Ta2O5 Formula Weight: 441.89 Physical Form: Powder Density: 8.2 Melting Point: 1800° Merck Reference: 14,9056 MDL No.: MFCD00011254

Formula: O₅Ta₂
MW: 441.89 g/mol
Melting Pt: 1800 °C
Density: 8.2
Storage Temperature: Ambient
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,9056

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR