Strontium bromide hexahydrate ≥95%

Supplier: Thermo Scientific Chemicals
Warning

72105-22 72105-36
CAAA72105-22EA 66.68 CAD
CAAA72105-22 CAAA72105-36
Strontium bromide hexahydrate ≥95%
Strontium bromide hexahydrate

MDL: MFCD00150361 Notes: m.p. 88°C (rapidly heated) , 180°C -6H2O Soluble in water, alcohol


Purity: 95% CAS No.: 7789-53-9 Molecular Formula: Br2Sr.6H2O Formula Weight: 355.53 (247.44anhy) Physical Form: Crystalline Density: 2.386 Appearance: White Odor: Odorless Solubility: Soluble in water, alcohol Merck Reference: 14,8837 MDL No.: MFCD00150361

Formula: Br₂Sr·6H₂O
MW: 355.52
Density: 2.386
Storage Temperature: Ambient
MDL Number: MFCD00150361
CAS Number: 7789-53-9
EINECS: 233-969-5
Merck Index: 14,8837

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR