Dioctyltin oxide

Supplier: Thermo Scientific Chemicals
Warning

71193-14 71193-22 71193-36
CAAA71193-14EA 47.87 CAD
CAAA71193-14 CAAA71193-22 CAAA71193-36
Dioctyltin oxide
Dioctyltin oxide

MDL: MFCD00013839


CAS No.: 870-08-6 Molecular Formula: (C8H17)2SnO Formula Weight: 361.14 Physical Form: Powder Appearance: White Melting Point: 245-248° dec. MDL No.: MFCD00013839

Formula: C₁₆H₃₄OSn
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 245…248 °C (dec.)
Density: 1,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013839
CAS Number: 870-08-6
EINECS: 212-791-1
UN: 3146
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR