Dimethyltin oxide
Supplier: Thermo Scientific Chemicals
|
Danger
|
71164-04
71164-09
71164-18
CAAA71164-04EA
58.14
CAD
CAAA71164-04
CAAA71164-09
CAAA71164-18
Dimethyltin oxide
Dimethyltin oxide
MDL: MFCD00013838
CAS No.: 2273-45-2
Molecular Formula: (CH3)2SnO
Formula Weight: 166.77
Physical Form: Powder
Melting Point: dec.
UN No.: 3146
MDL No.: MFCD00013838
Formula:
C₂H₆OSn Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00013838 CAS Number: 2273-45-2 EINECS: 218-881-7 UN: 3146 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...